1,4-Dioxane ≥99.0% (by GC) stabilised
: TCI
:123-91-1
:
:C₄H₈O₂
:MFCD00006571
:88.11 g/mol
:14,03300
:101°C
:11.8 °C
:1,03 g/cm³ (20 °C)
:12°C
:Ambient
:1165
:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
:204-661-8
:GHS02
:GHS07
:GHS08
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %