Triheptylamine ≥98.0% (by GC, titration analysis) for ion pair chromatography
TCIAI0361-25ML
: TCI
:
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
:
:155°C
:0,8119 g/cm³ (15 °C)
:311.6 g/mol
:219-316-7
:GHS07
:Ambient
:2411-36-1
:C₂₁H₄₅N
:MFCD00041936