Acrylonitrile ≥99.0% (by GC) stabilised
: TCI
:107-13-1
:
:C₃H₃N
:MFCD00001927
:53.06 g/mol
:14,00131
:79°C
:–84°C
:0,7951 g/cm³ (20 °C)
:–6°C
:1093
:3,I
:Stabilised with 4-methoxyphenol (MEHQ)
:203-466-5
:GHS02
:GHS05
:GHS06
:GHS08
:GHS09
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %