

Tris(dimethylamino)phosphine ≥96.0% (by GC) (may contain precipitate)
Supplier: TCI
Certificates
About this item
CAS Number:1608-26-0
:
Formula:C₆H₁₈N₃P
MDL Number:MFCD00008301
:163.2 g/mol
Boiling Pt:163°C
Melting Pt:–44 °C
Density:0,8962 g/cm³ (20 °C)
Flash Pt:26°C
Storage Temperature:Refrigerator
EINECS:216-534-4
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 96.0 %