Order Entry
Norway
ContactUsLinkComponent
Benzalkonium chloride 50% in water
Benzalkonium chloride 50% in water
Supplier:  TCI
Certificates
undefined
Benzalkonium chloride 50% in water
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Concentration (Nonaqueous Titration):
    50.0 to 55.0 w/w%

Specification Test Results