Order Entry
Norway
ContactUsLinkComponent
 
Tetramethylammonium hydroxide ∼25% in water
 :  TCI
 
undefined
Tetramethylammonium hydroxide ∼25% in water
 :  TCI

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Concentration (Neutralization titration):
    24.0 to 26.4 w/w%