
2,3,4-Trifluorophenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAT2574-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (Neutralization titration):97.0 to 112.0 %
Specification Test Results
About this item
:
:175.9 g/mol
:GHS07
CAS Number:226396-32-3
Formula:C₆H₄BF₃O₂
MDL Number:MFCD01863168