
N,N-Diethyl-m-toluidine ≥99.0% (by GC) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:91-67-8
:
Formula:C₁₁H₁₇N
MDL Number:MFCD00035795
Boiling Pt:107°C
Density:0,9051 g/cm³ (15 °C)
Flash Pt:104°C
:163.26 g/mol
EINECS:202-089-3
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 99.0 %
- Refractive index n20/D:1.5350 to 1.5380
- Hydrogendioxide detection test (Peroxidase method):to pass test