
Barbituric acid ≥99.0% (by titrimetric analysis) for colorimetric analysis of furfural derivatives
Catalog # TCIAB1063-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White powder to crystal
- Purity (Neutralization titration):min. 99.0 %
- Solubility in hot Water:clear and transparency
Specification Test Results
About this item
:
:128.09 g/mol
Merck Index:14,00963
Boiling Pt:441 °C (1013 hPa)
Melting Pt:248°C
Storage Temperature:Ambient
EINECS:200-658-0
:GHS07
CAS Number:67-52-7
Formula:C₄H₄N₂O₃
MDL Number:MFCD00006666