
2,3-Dimethoxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:40972-86-9
:
Formula:C₈H₁₁BO₄
MDL Number:MFCD02683112
:181.98 g/mol
Melting Pt:68°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 111.0 %