Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fmoc-Met-OH
Fmoc-Met-OH
Fmoc-Met-OH
04-12-1003-100
:
Novabiochem (Part of Merck)
:
Fmoc-Met-OH
04-12-1003-100
:
Novabiochem (Part of Merck)
:
Pack type:
Plastic bottle for solids
Pk:
100 g
Seq:
3401
:
:
371.46 g/mol
:
276-258-5
:
71989-28-1
:
C₂₀H₂₁NO₄S
:
MFCD00037134