Order Entry
Norway
ContactUsLinkComponent
 
3-Chloro-2,4,5,6-tetrafluorobenzotrifluoride ≥98.0% (by GC)
  TCIAC1727-25G
 :  TCI
 :  
undefined
3-Chloro-2,4,5,6-tetrafluorobenzotrifluoride ≥98.0% (by GC)
  TCIAC1727-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :137°C
 :GHS07
 :Refrigerator
 :4284-09-7
 :C₇ClF₇
 :MFCD03094143