Order Entry
Norway
Orders LinkContactUsLinkComponent
 

Phloretin ≥98%

Phloretin ≥98%
Certificates
Phloretin ≥98%
Phloretin ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 60-82-2 Molecular Formula: C15H14O5 Formula Weight: 274.28 Storage Temperature: 2°C to 8°C Melting Point: ca 263° dec. Beilstein Reference: 1887240 Merck Reference: 14,7326 MDL No.: MFCD00002288

  • CAS Number:60-82-2
  • Formula:C₁₅H₁₄O₅
  • MDL Number:MFCD00002288
  • Molecular Weight:274.28 g/mol
  • Merck Index:14,7326
  • Melting Pt:∼ 263 °C (dec.)
  • EINECS:200-488-7
Product Family Options

Product Information