2-Amino-3,5-dibromopyrazine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:24241-18-7
:
Formula:C₄H₃Br₂N₃
MDL Number:MFCD00673150
:252.90 g/mol
:GHS07
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:113.0 to 117.0 deg-C