
(S)-(+)-3-Hydroxytetrahydrofuran ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:86087-23-2
:
Formula:C₄H₈O₂
MDL Number:MFCD00064327
:88.11 g/mol
Boiling Pt:80°C
Flash Pt:79°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Optical purity (GC):min. 98.0 ee%