
2-Naphthonitrile ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:613-46-7
:
Formula:C₁₁H₇N
MDL Number:MFCD00016807
:153.18 g/mol
Boiling Pt:156°C
EINECS:210-344-5
UN:3439
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:65.0 to 69.0 deg-C
- Solubility in Methanol:almost transparency