Order Entry
Norway
ContactUsLinkComponent
8-Hydroxyquinoline-5-sulphonic acid monohydrate ≥98.0% (by titrimetric analysis)
8-Hydroxyquinoline-5-sulphonic acid monohydrate ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
8-Hydroxyquinoline-5-sulphonic acid monohydrate ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %
  • Water:
    7.0 to 7.8 %

Specification Test Results