Order Entry
Norway
ContactUsLinkComponent
 
 
Acetonyltriphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
  TCIAA1305-25G
 :  TCI
 :  
undefined
 
Acetonyltriphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
  TCIAA1305-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Solubility in hot Water:
    almost transparency

 

 

 :
 :354.82 g/mol
 :214-974-1
 :GHS07
 :1235-21-8
 :C₂₁H₂₀ClOP
 :MFCD00011813