Order Entry
Norway
ContactUsLinkComponent
 
 
Spermine tetrahydrochloride ≥98.0% (by total nitrogen and titration analysis)
 :  TCI
 
undefined
 
Spermine tetrahydrochloride ≥98.0% (by total nitrogen and titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Solubility in Water:
    almost transparency