
3-Amino-5-methylthio-1H-1,2,4-triazole ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:45534-08-5
:
Formula:C₃H₆N₄S
MDL Number:MFCD00005232
EINECS:256-242-4
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:134.0 to 139.0 deg-C
- Solubility in Water:very faint turbidity
Specification Test Results
Frequently Bought Together