γ-Cyclodextrine ≥99.0% (by HPLC)
: TCI
:17465-86-0
:
:C₄₈H₈₀O₄₀
:MFCD00009595
:1297.1248 g/mol
:14,02718
:241-482-4
- Appearance:White powder to crystal
- Purity (HPLC):min. 99.0 area%
- Specific rotation [a]20/D:+176.0 to +180.0 deg (C=1, H2O) (calcd.on anh.substance)
- Water:max. 12.0 %