Glycine methyl ester hydrochloride (H-Gly-OMe.HCl) ≥97.0% (by HPLC, total nitrogen)
Leverandør: TCI
Sertifikat
Om dette produktet
CAS-nummer:5680-79-5
:
Formel:C₃H₈ClNO₂
MDL-nummer:MFCD00012870
:125.56 g/mol
EINECS:227-139-1
Spesifikasjonstestresultater
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Solubility in Water:almost transparency