2,5-Diphenyl-p-benzoquinone ≥99.0% (by GC)
Catalog # TCIAD1166-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:216.0 to 220.0 deg-C
- Solubility in hot Toluene:almost transparency
Specification Test Results
About this item
:
:260.29 g/mol
EINECS:212-680-8
CAS Number:844-51-9
Formula:C₁₈H₁₂O₂
MDL Number:MFCD00001600