
4-Hydroxyanilinium chloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:51-78-5
:
Formula:C₆H₈ClNO
MDL Number:MFCD00012996
:145.59 g/mol
Merck Index:14,00462
EINECS:200-122-6
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Solubility in Water:almost transparency