Order Entry
Norway
ContactUsLinkComponent
2,5-Dichlorothiophene ≥97.0% (by GC)
Flammable
Toxic
2,5-Dichlorothiophene ≥97.0% (by GC)
Catalog # TCIAD0250-25G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Toxic
2,5-Dichlorothiophene ≥97.0% (by GC)
Catalog # TCIAD0250-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pakketype:
    Glass bottle
  • Fp:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :153.03 g/mol
Boiling Pt:162°C
Melting Pt:–40 °C
Density:1,4473 g/cm³ (20 °C)
Flash Pt:59°C
Storage Temperature:Kjøleskap
EINECS:221-638-8
UN:2929
ADR:6.1,II
 :GHS02
 :GHS06
CAS Number:3172-52-9
Formula:C₄H₂Cl₂S
MDL Number:MFCD00005423