2-Isopropylnaphthalene ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2027-17-0
:
Formula:C₁₃H₁₄
MDL Number:MFCD00014322
:170.25 g/mol
Boiling Pt:268°C
Melting Pt:11 °C
Density:0,9762 g/cm³ (20 °C)
Flash Pt:122°C
EINECS:217-976-0
ADR:9,III
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %