Order Entry
Norway
ContactUsLinkComponent
 
 
 
1,5-Dinitronaphthalene ≥98.0% (by GC)
 :  TCI
 
undefined
 
 
1,5-Dinitronaphthalene ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    214.0 to 219.0 deg-C
  • Solubility in hot THF:
    almost transparency