1-Nitronaphthalene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:86-57-7
:
Formula:C₁₀H₇NO₂
MDL Number:MFCD00003913
:173.17 g/mol
Merck Index:14,06613
Boiling Pt:304 °C (1013 hPa)
Melting Pt:59…61 °C
Density:1,331 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-684-5
UN:2538
ADR:4.1,III
:GHS02
:GHS06
:GHS09
Specification Test Results
- Appearance:White to Amber powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:56.0 to 59.0 deg-C
- Solubility in hot Methanol:almost transparency