
5-Aminoindan ≥98.0% (by GC, titration analysis)
TCIAA0991-10G
: TCI
:
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:34.0 to 38.0 deg-C
:
:133.19 g/mol
:147°C
:35 °C
:Refrigerator
:246-241-7
:6.1,III
:GHS07
:24425-40-9
:C₉H₁₁N
:MFCD00003803