Order Entry
Norway
Orders LinkContactUsLinkComponent
Orthoboric acid

Orthoboric acid

Health
Supplier:   G-Biosciences
Orthoboric acid
Supplier:  G-Biosciences
Certificates
Orthoboric acid

2 Options Available Below

Product Details & Documents
Synonyms:  Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
  • CAS Number:10043-35-3
  • Formula:B(OH)₃
  • MDL Number:MFCD00011337
  • Molecular Weight:61.83 g/mol
  • Merck Index:13,01326
  • Boiling Pt:185 °C (1013 hPa)
  • Melting Pt:160 °C
  • Density:1.44 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:233-139-2
  • REACH:01-2119486683-25
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - kr 695,00
    kr 695,00
    1 * 500 g
       kr 695,00

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - kr 1 185,00
    kr 1 185,00
    1 * 1 kg
       kr 1 185,00