
2-Methyl-2-thiazoline ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2346-00-1
:
Formula:C₄H₇NS
MDL Number:MFCD00005314
:101.17 g/mol
Boiling Pt:145°C
Flash Pt:37°C
Storage Temperature:Refrigerator
EINECS:219-071-6
UN:1993
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %