
Acetamidine hydrochloride ≥95.0% (by titrimetric analysis)
: TCI
:124-42-5
:
:C₂H₇ClN₂
:MFCD00013016
:94.54 g/mol
:14,00044
:204-700-9
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 95.0 % (after deducting NH4Cl)
- Purity (Argentmetric Titration):min. 95.0 % (after deducting NH4Cl)
- Melting point:166.0 to 176.0 deg-C
- Ammonium chloride:max. 5.0 %