Bis(diphenylphosphino)methane ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2071-20-7
:
Formula:C₂₅H₂₂P₂
MDL Number:MFCD00003537
:384.4 g/mol
EINECS:218-194-2
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Iodometric back titration):min. 97.0 %
- Melting point:118.0 to 122.0 deg-C
- Solubility in Toluene:almost transparency