Bromocyclohexane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:108-85-0
:
Formula:C₆H₁₁Br
MDL Number:MFCD00003819
:163.06 g/mol
Merck Index:14,02730
Boiling Pt:165°C
Melting Pt:–56.5 °C
Density:1,33 g/cm³ (20 °C)
Flash Pt:62°C
Storage Temperature:Ambient
EINECS:203-622-2
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %