

Benzyl benzoate ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:120-51-4
:
Formula:C₁₄H₁₂O₂
MDL Number:MFCD00003075
:212.25 g/mol
Merck Index:14,01127
Boiling Pt:323 °C (1013 hPa)
Melting Pt:17…20 °C
Density:1,118 g/cm³ (20 °C)
Flash Pt:158°C (closed cup)
Storage Temperature:Ambient
EINECS:204-402-9
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
- Density (20deg.C):1.1160 to 1.1210 g/mL