Order Entry
Norway
ContactUsLinkComponent
 
 
 
2,6-Di-tert-butyl-4-ethylphenol ≥98.0% (by GC)
 :  TCI
 
undefined
 
 
2,6-Di-tert-butyl-4-ethylphenol ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    43.0 to 47.0 deg-C
  • Solubility in Methanol:
    almost transparency