Order Entry
Norway
ContactUsLinkComponent
 
 
2-Chloromethyl-p-xylene ≥98.0% (by GC, titration analysis)
  TCIAD2716-25G
 :  TCI
 :  
undefined
 
2-Chloromethyl-p-xylene ≥98.0% (by GC, titration analysis)
  TCIAD2716-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %

 

 

 :
 :154.64 g/mol
 :121°C
 :1,0678 g/cm³ (20 °C)
 :65°C
 :212-529-6
 :3265
 :8,III
 :GHS05
 :824-45-3
 :C₉H₁₁Cl
 :MFCD00000902