
2,5-Dichloro-p-xylene ≥97.0% (by GC)
Catalog # TCIAD0431-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:66.0 to 71.0 deg-C
Specification Test Results
About this item
:
:175.06 g/mol
Boiling Pt:108°C
Melting Pt:71 °C
Density:1,39 g/cm³ (20 °C)
EINECS:214-387-0
:GHS07
CAS Number:1124-05-6
Formula:C₈H₈Cl₂
MDL Number:MFCD00000610