

4-Fluoro-3-nitrobenzotrifluoride ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:367-86-2
:
Formula:C₇H₃F₄NO₂
MDL Number:MFCD00007059
:209.1 g/mol
Boiling Pt:208°C
Density:1,49 g/cm³ (20 °C)
Flash Pt:33°C
Storage Temperature:Ambient
EINECS:206-702-5
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Purity (GC):min. 95.0 %