
(1-Bromoethyl)benzene ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:585-71-7
:
Formula:C₆H₅CHBrCH₃
MDL Number:MFCD00000139
:185.06 g/mol
Boiling Pt:200°C
Melting Pt:50 °C
Density:1,3605 g/cm³ (20 °C)
Flash Pt:96°C
Storage Temperature:Refrigerator
EINECS:209-560-2
ADR:8,II
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %