
(±)-1-Phenyl-2-propyn-1-ol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4187-87-5
:
Formula:C₉H₈O
MDL Number:MFCD00021860
:132.16 g/mol
Boiling Pt:115°C
EINECS:224-064-6
:GHS07
Specification Test Results
- Appearance:White or Colorless to Light orange to Yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %