
1-Butyl-3-methylpyridinium chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:125652-55-3
:
Formula:C₁₀H₁₆ClN
MDL Number:MFCD08458942
:185.7 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %