
Methyl indole-3-carboxylate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:942-24-5
:
Formula:C₁₀H₉NO₂
MDL Number:MFCD00189407
:175.19 g/mol
:GHS07
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:148.0 to 152.0 deg-C