Dicyclopropyl ketone ≥95.0% (by GC)
Catalog # TCIAD1758-25ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Purity (GC):min. 95.0 %
Specification Test Results
About this item
:
:110.16 g/mol
Boiling Pt:67°C
Flash Pt:39°C
EINECS:214-331-5
UN:1224
ADR:3,III
:GHS02
:GHS05
:GHS07
CAS Number:1121-37-5
Formula:C₇H₁₀O
MDL Number:MFCD00001276