Order Entry
Norway
ContactUsLinkComponent
N-Butylaniline ≥98.0% (by GC, titration analysis)
Irritant
Health
N-Butylaniline ≥98.0% (by GC, titration analysis)
Catalog # TCIAB0711-25ML
Supplier:  TCI
CAS Number:  
undefined
Irritant
Health
N-Butylaniline ≥98.0% (by GC, titration analysis)
Catalog # TCIAB0711-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :149.24 g/mol
Boiling Pt:240°C
Melting Pt:–14.4 °C
Density:0,9342 g/cm³ (20 °C)
Flash Pt:105°C
EINECS:214-425-6
UN:2738
ADR:6.1,II
 :GHS07
 :GHS08
CAS Number:1126-78-9
Formula:(C₆H₅)NHCH₂CH₂CH₂CH₃
MDL Number:MFCD00035798