kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
3-Fluorothiophenol
3-Fluorothiophenol ≥97.0% (by GC)
3-Fluorothiophenol ≥97.0% (by GC)
:
TCI
3-Fluorothiophenol ≥97.0% (by GC)
:
TCI
:
2557-77-9
:
:
C₆H₅FS
:
MFCD00040227
:
128.17 g/mol
:
156°C
:
1,18 g/cm³ (30 °C)
:
219-876-2
:
3336
:
3,III
:
GHS02
:
GHS07
Appearance:
Colorless to Almost colorless clear liquid
Purity (GC):
min. 97.0 %