
2,3,4,5-Tetrafluorobenzoic acid ≥98.0% (by HPLC, titration analysis)
: TCI
:1201-31-6
:
:C₇H₂F₄O₂
:MFCD00009613
:194.09 g/mol
:135 - 137 °C (23 -rr)
:92…92.5 °C
:GHS07
:Ambient
- Appearance:White to Gray to Red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:86.0 to 89.0 deg-C
- Solubility in Methanol:almost transparency