Order Entry
Norway
ContactUsLinkComponent
2,5-Dichlorophenol ≥98.0% (by GC, titration analysis)
Irritant
Health
Environment
2,5-Dichlorophenol ≥98.0% (by GC, titration analysis)
Catalog # TCIAD0393-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Health
Environment
2,5-Dichlorophenol ≥98.0% (by GC, titration analysis)
Catalog # TCIAD0393-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light red to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    55.0 to 58.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :163 g/mol
Boiling Pt:211°C
Melting Pt:60…61 °C
Storage Temperature:Ambient
EINECS:209-520-4
UN:2020
ADR:6.1,III
 :GHS07
 :GHS08
 :GHS09
CAS Number:583-78-8
Formula:C₆H₄Cl₂O
MDL Number:MFCD00002174