


1-Chloro-3-nitrobenzene ≥99.0% (by GC)
: TCI
:121-73-3
:
:C₆H₄ClNO₂
:MFCD00007202
:157.56 g/mol
:14,02151
:236°C
:46 °C
:1,343 g/cm³ (20 °C)
:Ambient
:204-496-1
:1578
:6.1,II
:GHS06
:GHS08
:GHS09
- Appearance:White or Colorless to Yellow powder to lump to clear liquid
- Purity (GC):min. 99.0 %
- Melting point:44.0 to 48.0 deg-C
- Solubility in Methanol:almost transparency