
2,5-Dichlorobenzaldehyde ≥98.0% (by GC)
TCIAD2814-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:55.0 to 59.0 deg-C
:
:233°C
:228-832-1
:8,III
:175.01 g/mol
:1759
:GHS05
:6361-23-5
:C₇H₄Cl₂O
:MFCD00156140